Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dihydrolipoamide Dehydrogenase/DLD Antibody (OTI6D5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270574AF647
Description
Dihydrolipoamide Dehydrogenase/DLD Monoclonal specifically detects Dihydrolipoamide Dehydrogenase/DLD in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Dihydrolipoamide Dehydrogenase/DLD | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DLD | |
Full length recombinant protein of human Dihydrolipoamide Dehydrogenase/DLD (NP_000099) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI6D5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, lipoamide dehydrogenase, lipoamide reductase, lipoyl dehydrogenase | |
Mouse | |
Affinity Purified | |
Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience | |
1738.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction