Learn More
Dihydrotestosterone Polyclonal Antibody

Description
Dihydrotestosterone Polyclonal Antibody for ELISA
Specifications
Specifications
| Antigen | Dihydrotestosterone |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 10.98 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Andractim; Androlone; Androstanolone; C19H30O2; DHT; STANOLONE |
| Host Species | Sheep |
| Immunogen | Dihydrotestosterone(3)-BTG. |
| Purification Method | Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.