Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutyl phthalate, 99%, Thermo Scientific™

Catalog No. AAA1705936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1705936 500 g
AAA1705922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1705936 Supplier Thermo Scientific Chemicals Supplier No. A1705936
Only null left
Add to Cart

Chemical Identifiers

CAS 84-69-5
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD00026480
InChI Key MGWAVDBGNNKXQV-UHFFFAOYSA-N
Synonym diisobutyl phthalate, palatinol ic, dibp, isobutyl phthalate, hexaplas m/1b, kodaflex dibp, di-iso-butyl phthalate, phthalic acid, diisobutyl ester, phthalic acid diisobutyl ester, di i-butyl phthalate
PubChem CID 6782
ChEBI CHEBI:79053
IUPAC Name 1,2-bis(2-methylpropyl) benzene-1,2-dicarboxylate
SMILES CC(C)COC(=O)C1=CC=CC=C1C(=O)OCC(C)C

Specifications

Melting Point -64°C
Density 1.039
Boiling Point 327°C
Flash Point 169°C (336°F)
Refractive Index 1.49
Quantity 500 g
Beilstein 2054802
Formula Weight 278.35
Percent Purity 99%
Chemical Name or Material Diisobutyl phthalate
Hazard Category H360Df
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 201-553-2
RTECSNumber TI1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.