Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylaluminum hydride, 25% w/w in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. aa42592ad
Click to view available options
Quantity:
50 mL
200 mL
800 mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928

Specifications

Density 0.701 g/mL
Packaging ChemSeal™ bottles
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 50 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
Hazard Category H225-H250-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H224-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.