Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diisobutylaluminum hydride, 25% w/w in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
50 mL
200 mL
800 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Specifications
| Density | 0.701 g/mL |
| Packaging | ChemSeal™ bottles |
| Linear Formula | [(CH3)2CHCH2]2AlH |
| Quantity | 800 mL |
| UN Number | UN3399 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 142.22 |
| Physical Form | Liquid |
| Chemical Name or Material | Diisobutylaluminum hydride |
Safety and Handling
| Hazard Category | H225-H250-H260-H304-H314-H335-H336-H361f-H373 |
| Hazard Statement | GHS H Statement H224-H260-H304-H361-H373-H314-H318-H336 Extremely flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure. Causes severe skin burns and eye damage. Causes serious eye damage. May cause drowsiness or dizziness. |
| Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction