Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Diisobutylaluminum Hydride Solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
800 mL
Description
Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.
Specifications
Specifications
| CAS | 1191-15-7 |
| Density | 0.731 g/mL at 25°C |
| Molecular Formula | C8H19Al |
| Linear Formula | [(CH3)2CHCH2]2AlH |
| MDL Number | MFCD00008928 |
| Quantity | 800 mL |
| Synonym | DIBAL; DIBAL-H |
| Molecular Weight (g/mol) | 142.22 g/mol |
| Biological Activity | Central Nervous System |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction