Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropanolamine, 98+%, sum of isomers

Catalog No. AAA184510B
Click to view available options
Quantity:
250 g
1000 g

CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol

Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.

Solubility
Soluble in water 20°C 870g/L. Miscible in alcohol.

Notes
Air Sensitive & Hygroscopic. Store away from oxidizing agents, air, water/moisture and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-97-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00004531
InChI Key LVTYICIALWPMFW-UHFFFAOYNA-N
Synonym diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
PubChem CID 8086
IUPAC Name 1-[(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(O)CNCC(C)O

Specifications

Melting Point 38°C to 45°C
Density 1.004
Boiling Point 249°C to 250°C
Flash Point 130°C (266°F)
Odor Amine-like
Quantity 1000 g
Beilstein 605363
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,3195
Solubility Information Soluble in water 20°C 870g/L. Miscible in alcohol.
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material Diisopropanolamine, sum of isomers
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 203-820-9
RTECSNumber UB6600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.