Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropanolamine, 99%

Catalog No. p-113468
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-97-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00004531
InChI Key LVTYICIALWPMFW-UHFFFAOYNA-N
Synonym diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
PubChem CID 8086
IUPAC Name 1-[(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(O)CNCC(C)O

Specifications

Melting Point 33°C
Color Colorless to Yellow
Density 1.0038g/mL
Boiling Point 249°C to 250°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (Total base)
Packaging Glass bottle
Refractive Index 1.4615 to 1.4635
Linear Formula (CH3CHOHCH2)2NH
Quantity 500 g
Specific Gravity 1.0038
Merck Index 15, 3214
Solubility Information Solubility in water: 870g/L (20°C)
Viscosity 84.19 mPa.s (60°C)
Formula Weight 133.19
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Diisopropanolamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 203-820-9
RTECSNumber UB6600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.