Learn More
Diisopropyl azodicarboxylate, 94%
Description
Diisopropyl azodicarboxylate is used as an important reagent in the production of many organic compounds. It is used in association with triphenyl phosphine in Mitsunobu reaction of alcohols, acids and amides. It acts as reactant in the preparation of chromenes resembling classical cannabinoids, norbornene-based guanidine-rich polymers and acceptor-donor-acceptor organic dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diisopropyl azodicarboxylate is used as an important reagent in the production of many organic compounds. It is used in association with triphenyl phosphine in Mitsunobu reaction of alcohols, acids and amides. It acts as reactant in the preparation of chromenes resembling classical cannabinoids, norbornene-based guanidine-rich polymers and acceptor-donor-acceptor organic dyes.
Solubility
Immiscible with water.
Notes
Store in cool place. Light sensitive. Incompatible with strong oxidizing agents, strong bases, alcohols and metallic salts. Decomposes violently at or above: 100 0C.
Specifications
Specifications
| Melting Point | 3°C to 5°C |
| Density | 1.027 |
| Boiling Point | 74°C to 75°C (0.3 mmHg) |
| Flash Point | 160°C (320°F) |
| Odor | Pungent |
| Refractive Index | 1.42 |
| Quantity | 25 g |
| UN Number | UN3082 |
| Beilstein | 1912326 |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335-H373 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 219-502-8 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.