Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl D-(-)-Tartrate 98.0+%, TCI America™
SDP

Catalog No. T1387250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1387250G 250 g
T138725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1387250G Supplier TCI America Supplier No. T1387250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62961-64-2
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00008876
InChI Key XEBCWEDRGPSHQH-YUMQZZPRSA-N
Synonym diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
PubChem CID 112972
IUPAC Name dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O

Specifications

Color Yellow
Quantity 250 g
Formula Weight 234.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl D-(-)-Tartrate
EINECSNumber (2)-1458
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.