Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl fluorophosphate, 99%

Catalog No. ac115230010
Click to view available options
Quantity:
1 g

CAS: 55-91-4 | C6H14FO3P | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-91-4
Molecular Formula C6H14FO3P
Molecular Weight (g/mol) 184.15
InChI Key MUCZHBLJLSDCSD-UHFFFAOYSA-N
Synonym isoflurophate, diisopropyl fluorophosphate, diisopropylfluorophosphate, fluostigmine, isofluorphate, neoglaucit, diflupyl, dyflos, isoflurophosphate, diflurophate
PubChem CID 5936
ChEBI CHEBI:17941
IUPAC Name 2-[fluoro(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(=O)(OC(C)C)F

Specifications

Melting Point -82.0°C
Density 1.0550g/mL
Boiling Point 62.0°C (9.0 mmHg)
Assay Percent Range 98.5% min. (GC)
Packaging Ampoule
Linear Formula [(CH3)2CHO]2P(O)F
Quantity 1 g
Specific Gravity 1.055
Merck Index 15,5222
Solubility Information Solubility in water: 14.5g/L in water (25°C)- decomposes. Other solubilities: soluble in vegetable oils,alcohols and ether,not very soluble in mineral oils
Formula Weight 184.15
Percent Purity 99%
Chemical Name or Material Diisopropyl fluorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 200-247-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.