Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl phosphite, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
1809-20-7
C6H14O3P-
165.149
MFCD00117905
IWEDBEZKWHRUJX-UHFFFAOYSA-N
diisopropylphosphite,acmc-1bsaz,phosphorous acid diisopropyl anion,phosphorous acid, bis 1-methylethyl ester
21884976
dipropan-2-yl phosphite
CC(C)OP([O-])OC(C)C
1809-20-7
C6H14O3P-
165.149
MFCD00117905
IWEDBEZKWHRUJX-UHFFFAOYSA-N
diisopropylphosphite,acmc-1bsaz,phosphorous acid diisopropyl anion,phosphorous acid, bis 1-methylethyl ester
21884976
dipropan-2-yl phosphite
CC(C)OP([O-])OC(C)C
Applications
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

Solubility
Soluble in water, chloroform.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Flash Point 112°C (234°F)
Boiling Point 72°C to 75°C (10mmHg)
Quantity 25g
Linear Formula [(CH3)2CHO]2P(O)H
Formula Weight 166.16
Percent Purity 98%
Chemical Name or Material Diisopropyl phosphite

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : SZ7660000

TSCA : TSCA