Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diisopropyl phosphite, 98%
Click to view available options
Quantity:
25 g
100 g
Description
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.
Solubility
Soluble in water, chloroform.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.
Solubility
Soluble in water, chloroform.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications
Specifications
| Density | 0.99 |
| Boiling Point | 72°C to 75°C (10 mmHg) |
| Flash Point | 112°C (234°F) |
| Refractive Index | 1.4075 |
| Linear Formula | [(CH3)2CHO]2P(O)H |
| Quantity | 100 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water,chloroform. |
| Formula Weight | 166.16 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H332-H315-H319-H335 Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 217-317-7 |
| RTECSNumber | SZ7660000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction