Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylamine, 99%, Thermo Scientific Chemicals

Catalog No. p-2691358
Click to view available options
Quantity:
1 L
2.5 L

Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
PubChem CID 7912
IUPAC Name N-propan-2-ylpropan-2-amine
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Color Colorless
Density 0.7160g/mL
Boiling Point 84.0°C
Flash Point −17°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C6H15N
Refractive Index 1.3910 to 1.3930
Linear Formula [(CH3)2CH]2NH
Quantity 1 L
UN Number 1158
Beilstein 04, 154
Specific Gravity 0.716
Merck Index 15, 3215
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol, soluble in most common organic solvents
Molecular Weight (g/mol) 101.19
Viscosity 0.4 mPa.s (20°C)
Vapor Pressure 95mbar at 20°C
Formula Weight 101.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diisopropylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 203-558-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.