Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Diltiazem hydrochloride
GREENER_CHOICESDP

Catalog No. 06851G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-851-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-851-G Supplier R&D Systems™ Supplier No. 0685/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CaV1.x blocker

Antihypertensive and cardioprotective agent; an inhibitor of L-type Ca2+ channels.

Chemical Identifiers

CAS 33286-22-5
Molecular Formula C22H27ClN2O4S

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 450.98
Purity 98%
Molecular Weight (g/mol) 450.978
Chemical Name or Material Diltiazem hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.