Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience EasySpiral Counting Grid
SDP

Catalog No. 16948986 Shop All Interscience Products
Click to view available options
Diameter (Metric):
90 mm
150 mm

Spiral counting grid for use with EasySpiral platers.

  • Spiral counting grid
  • For use with EasySpiral platers
  • Available for 90 mm or 150 mm Petri dishes

Specifications

Product Type Counting Grids
Display Type None
Diameter (Metric) 150 mm
For Use With (Equipment) EasySpiral
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.