Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor

Accurately dispense media into suitable containers according to a set dilution using this gravimetric diluter which allows the use of two diluents.

$10640.00 - $14935.00

Specifications

For Use With Dry-Bags
Material Stainless Steel (Case)
Overall Length (Metric) 41.5 cm
Overall Width (Metric) 31 cm
Electrical Requirements 100/127 V 220/240 V 50/60 Hz
View More Specs

Products 2
Catalog Number Mfr. No. Includes Model Price Quantity & Availability  
Catalog Number Mfr. No. Includes Model Price Quantity & Availability  
DB4111A
View Documents
Thermo Scientific™
DB4100A/DUP
Simple Dispensing Arm DiluFlux Pro-3
Each for $10,640.00
Only null left
Add to Cart
 
DB4150A
View Documents
Thermo Scientific™
DB4150A
Robotic Dispensing Arm DiluFlux Pro-5
Each for $14,935.00
Only null left
Add to Cart
 
Description

Description

Accurately dispense broth medium into suitable containers for food testing using the Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor. DiluFlux Pro Dilutors are compact, easy-to-use, and designed to deliver precise quantities of diluents rapidly with the touch of a button, so that you can work more confidently and accurately. When combined with the Thermo Scientific™ Peristaltic Pump and Thermo Scientific™ Homogenizer, your lab bench will be equipped with a flexible automated culture media solution to increase your productivity.

Automate the dilution step of your Thermo Scientific™ Dry-Bags™ Enrichment Media, as well as other formats, for maximum efficiency. The easy-to-use DiluFlux Pro Gravimetric Dilutors allow the simultaneous use of two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted into the unit.

  • Intuitive: simply press two keys, tare and go, and the DiluFlux Pro Dilutor will automatically dilute your sample to the desired dilution factor
  • Accurate: featuring Watson-Marlow™ peristaltic pumps, high-tech flow regulators, and top of the line weighing platforms for trusted reproducibility and repeatability
  • Versatile: compatible with many prepared media formats, the easy-to clean, stainless steel DiluFlux Pro Dilutor is an ideal fit for any food testing laboratory
  • High-quality: the DiluFlux Pro Dilutors are ISO 7218, ISO 6887-1, FDA BAM-compliant and are made in France under ISO 9001:2008 certification

Not all products are available for sale in all territories. Please inquire.

Specifications

Specifications

Dry-Bags
41.5 cm
100/127 V 220/240 V 50/60 Hz
50/60 Hz
Stainless Steel (Case)
31 cm
100/240 V
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.