Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DIM-C-pPhOH
GREENER_CHOICESDP

Catalog No. 63775 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-775 5 mg
63-772-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-775 Supplier Tocris Bioscience™ Supplier No. 6377/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nur77 antagonist

DIM-C-pPhOH is a Nur77 (NR4A1) antagonist. Inhibits TGF-β induced cell migration of breast cancer cell lines. Promotes ROS/endoplasmic reticulum stress and proapoptotic pathways in pancreatic cancer cell lines. Mimics effects of Nur77 RNAi silencing.

Specifications

Chemical Name or Material 1,1-Bis(3'-indolyl)-1-(p-hydroxyphenyl)methane
CAS 151358-47-3
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C23H18N2O
Formula Weight Observed MW: 338.408
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.