Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimesitylfluoroborane 98.0+%, TCI America™
SDP

Catalog No. D38901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38901G 1 g
D38905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38901G Supplier TCI America Supplier No. D38901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 436-59-9
Molecular Formula C18H22BF
Molecular Weight (g/mol) 268.18
MDL Number MFCD00012203
InChI Key WZWGERGANZMXOM-UHFFFAOYSA-N
Synonym Dimesitylboron Fluoride
PubChem CID 3466040
IUPAC Name fluorobis(2,4,6-trimethylphenyl)borane
SMILES CC1=CC(C)=C(B(F)C2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 129°C
Quantity 1 g
UN Number 3261
Formula Weight 268.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dimesitylfluoroborane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.