Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dimethomorph Monoclonal Antibody (4A5)
GREENER_CHOICE

Catalog No. PIMA549703
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549703 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549703 Supplier Invitrogen™ Supplier No. MA549703
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Dimethomorph is a fungicide that belongs to the class of chemicals known as cyanoacetamides. It is primarily used to control various types of mildew and blight on crops such as grapes, potatoes, and vegetables. The mode of action of Dimethomorph is to inhibit the development of fungal cell walls, thereby preventing the growth and spread of the disease. It is a systemic fungicide, which means that it is absorbed by the plant and transported to all parts of the plant, providing long-lasting protection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dimethomorph
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4A5
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of DMM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.