Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxymethylphenylsilane 98.0+%, TCI America™
SDP

Catalog No. D2319100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2319100ML 100 mL
D231925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2319100ML Supplier TCI America Supplier No. D2319100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3027-21-2
Molecular Formula C9H14O2Si
Molecular Weight (g/mol) 182.29
MDL Number MFCD00008343
InChI Key CVQVSVBUMVSJES-UHFFFAOYSA-N
Synonym dimethoxymethylphenylsilane, dimethoxy methyl phenyl silane, methylphenyldimethoxysilane, phenylmethyldimethoxysilane, silane, dimethoxymethylphenyl, dimethoxyphenylmethylsilane, fenyl-dimethoxy-methylsilan, dimethoxy methyl phenylsilane, fenyl-dimethoxy-methylsilan czech, methylphenyldimethoxy silane
PubChem CID 18193
IUPAC Name dimethoxy(methyl)phenylsilane
SMILES CO[Si](C)(OC)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 199°C
Quantity 100 mL
Formula Weight 182.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxymethylphenylsilane
RTECSNumber VV3645000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.