Learn More
Dimethyl 5-aminoisophthalate, 98%
Description
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.
Solubility
Soluble in Chloroform.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 178°C to 182°C |
| Quantity | 25 g |
| Beilstein | 2116565 |
| Synonym | dimethyl 5-aminoisophthalate, 3,5-dicarbomethoxyaniline, 5-aminoisophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-amino-, dimethyl ester, 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate, dimethyl5-aminoisophthalate, 5-amino-isophthalic acid dimethyl ester, 5-amino-dimethyl iso-phthalate, 5-amino-dimethylisophthalate, dimethyl-5-aminoisophthalate |
| Solubility Information | Soluble in Chloroform. |
| Formula Weight | 209.2 |
| Percent Purity | 98% |
| Chemical Name or Material | Dimethyl 5-aminoisophthalate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-743-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.