Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyl 5-hydroxyisophthalate, 98%

Click to view available options
Quantity:
10g
50g
250g
13036-02-7
C10H10O5
210.185
MFCD00134367
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
dimethyl 5-hydroxyisophthalate,5-hydroxyisophthalic acid dimethyl ester,1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate,1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester,dimethyl5-hydroxyisophthalate,timtec-bb sbb000491,5-hydroxy-dimethylisophthalate,1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester,dimethyl-5-hydroxy-benzene-1,3-dicarbonate,methyl 5-hydroxy-3-methoxycarbonyl benzoate
83065
dimethyl 5-hydroxybenzene-1,3-dicarboxylate
COC(=O)C1=CC(=CC(=C1)O)C(=O)OC
13036-02-7
C10H10O5
210.185
MFCD00134367
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
dimethyl 5-hydroxyisophthalate,5-hydroxyisophthalic acid dimethyl ester,1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate,1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester,dimethyl5-hydroxyisophthalate,timtec-bb sbb000491,5-hydroxy-dimethylisophthalate,1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester,dimethyl-5-hydroxy-benzene-1,3-dicarbonate,methyl 5-hydroxy-3-methoxycarbonyl benzoate
83065
dimethyl 5-hydroxybenzene-1,3-dicarboxylate
COC(=O)C1=CC(=CC(=C1)O)C(=O)OC
Applications
Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Melting Point 162°C to 164°C
Quantity 10g
Beilstein 2650120
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material Dimethyl 5-hydroxyisophthalate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA