Alfa Aesar™ Dimethyl acetylenedicarboxylate, 95%

Manufacturer: Alfa Aesar™ 4513136

Quantity 500g

 View more versions of this product

Catalog No. AA4513136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 762-42-5
    Molecular Formula C6H6O4
    Linear Formula CH3O2CC≡CCO2CH3
    Formula Weight 142.11
    MDL Number MFCD00008456
    UN Number UN1760
    Beilstein 607063
    Synonym Acetylenedicarboxylic acid dimethyl ester
    Chemical Name or Material Dimethyl acetylenedicarboxylate

    Specifications

    Formula Weight 142.11
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula CH3O2CC≡CCO2CH3
    CAS 762-42-5
    Health Hazard 1 Danger
    TSCA TSCA
    Melting Point −18°C
    Boiling Point 95° to 98°C (19mmHg)
    MDL Number MFCD00008456
    UN Number UN1760
    Quantity 500g
    Sensitivity Light Sensitive
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Density 1.156g/mL
    Color colorless to pale yellow
    Synonym Acetylenedicarboxylic acid dimethyl ester
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Combustible liquid.
    Refractive Index 1.4470
    Physical Form Liquid
    Molecular Formula C6H6O4
    Beilstein 607063
    Flash Point 86°C (186°F)
    RTECS Number ES0175000
    Chemical Name or Material Dimethyl acetylenedicarboxylate
    Assay 95%

    GHS H Statement
    H314-H318-H302-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit