Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl acetylsuccinate, 96%

Catalog No. p-7028228
Click to view available options
Quantity:
100 g
500 g

CAS: 10420-33-4 | C8H12O5 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10420-33-4
Molecular Formula C8H12O5
Molecular Weight (g/mol) 188.18
MDL Number MFCD00008448
InChI Key XREKLQOUFWBSFH-UHFFFAOYNA-N
Synonym dimethyl acetylsuccinate, dimethyl 2-acetylsuccinate, 1,4-dimethyl 2-acetylbutanedioate, butanedioic acid, acetyl-, dimethyl ester, acetylsuccinic acid dimethyl ester, dmas, butanedioic acid, 2-acetyl-, 1,4-dimethyl ester, doimethyl acetylsuccinate, acmc-1bwle, dimethyl 2-acetylsuccite
PubChem CID 112039
IUPAC Name dimethyl 2-acetylbutanedioate
SMILES COC(=O)CC(C(C)=O)C(=O)OC

Specifications

Melting Point 30°C to 33°C
Density 1.16
Boiling Point 129°C to 134°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Formula Weight 188.18
Percent Purity 96%
Chemical Name or Material Dimethyl acetylsuccinate
EINECSNumber 233-897-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.