Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Adipimidate Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. A080625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A080625G 25 g
A08065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A080625G Supplier TCI America Supplier No. A080625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14620-72-5
Molecular Formula C8H18Cl2N2O2
Molecular Weight (g/mol) 245.144
MDL Number MFCD00012573
InChI Key IEUUDEWWMRQUDS-UHFFFAOYSA-N
Synonym dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride
PubChem CID 12876176
IUPAC Name dimethyl hexanediimidate;dihydrochloride
SMILES COC(=N)CCCCC(=N)OC.Cl.Cl

Specifications

Melting Point 215°C
Color White
Quantity 25 g
Formula Weight 245.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Adipimidate Dihydrochloride [Cross-linking Agent for Protein Research]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.