Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Benzylmalonate 95.0+%, TCI America™
SDP

Catalog No. D343225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D343225G 25 g
D34325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D343225G Supplier TCI America Supplier No. D343225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49769-78-0
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD01445471
InChI Key ZMYJSOIMFGAQRQ-UHFFFAOYSA-N
Synonym Benzylmalonic Acid Dimethyl Ester
PubChem CID 10900856
IUPAC Name 1,3-dimethyl 2-benzylpropanedioate
SMILES COC(=O)C(CC1=CC=CC=C1)C(=O)OC

Specifications

Color Colorless
Boiling Point 285°C
Quantity 25 g
Formula Weight 222.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Benzylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.