Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Bromoterephthalate 98.0+%, TCI America™
SDP

Catalog No. p-7128601
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 18643-86-2
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00060638
InChI Key VUMPFOPENBVFOF-UHFFFAOYSA-N
Synonym Bromoterephthalic Acid Dimethyl Ester
PubChem CID 87741
IUPAC Name dimethyl 2-bromobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)Br

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 273.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyl Bromoterephthalate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.