Learn More
Dimethyl carbonate, 99%
Description
Dimethyl carbonate is used as a solvent in organic synthesis and considered as a replacement for solvent like methyl ethyl ketone, tert-butyl acetate and parachlorobenzotrifluoride. It is involved as an intermediate in the preparation of diphenylcarbonate, which in turn is used as a key raw material for the synthesis of Bisphenol-A-polycarbonate. It is also used as a 'green' methylating agent involved in the methylation of aniline, phenols and carboxylic acids. It can be used as a fuel additive due to its high oxygen content. It also finds applications related to supercapacitors and lithium batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl carbonate is used as a solvent in organic synthesis and considered as a replacement for solvent like methyl ethyl ketone, tert-butyl acetate and parachlorobenzotrifluoride. It is involved as an intermediate in the preparation of diphenylcarbonate, which in turn is used as a key raw material for the synthesis of Bisphenol-A-polycarbonate. It is also used as a ′green′ methylating agent involved in the methylation of aniline, phenols and carboxylic acids. It can be used as a fuel additive due to its high oxygen content. It also finds applications related to supercapacitors and lithium batteries.
Solubility
Miscible with alcohol and ether. Immiscible with water.
Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | ∽4°C |
| Density | 1.070 |
| Boiling Point | 90°C |
| Flash Point | 16°C (61°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.368 |
| Quantity | 100 g |
| UN Number | UN1161 |
| Beilstein | 635821 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225 |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYL CARBONATE |
| EINECSNumber | 210-478-4 |
| RTECSNumber | FG0450000 |
| TSCA | Yes |
| storageNote1 | Moisture Sensitive |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.