Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Chloromalonate 98.0+%, TCI America™
SDP

Catalog No. D546725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D546725G 25 g
D54675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D546725G Supplier TCI America Supplier No. D546725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28868-76-0
Molecular Formula C5H7ClO4
Molecular Weight (g/mol) 166.56
MDL Number MFCD00042837
InChI Key LNBQBURECUEBKZ-UHFFFAOYSA-N
Synonym Chloromalonic Acid Dimethyl Ester
PubChem CID 120070
IUPAC Name 1,3-dimethyl 2-chloropropanedioate
SMILES COC(=O)C(Cl)C(=O)OC

Specifications

Color Yellow
Boiling Point 208°C
Quantity 25 g
UN Number 3265
Formula Weight 166.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Chloromalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.