Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Citraconate 99.0+%, TCI America™
SDP

Catalog No. C036425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 617-54-9
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00059306
InChI Key WQEXBUQDXKPVHR-SNAWJCMRSA-N
Synonym Citraconic Acid Dimethyl Ester
PubChem CID 5355715
IUPAC Name 1,4-dimethyl (2E)-2-methylbut-2-enedioate
SMILES COC(=O)\C=C(/C)C(=O)OC

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 g
Formula Weight 158.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Citraconate
RTECSNumber GE6700000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.