Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl diselenide, 99%

Catalog No. AA6811906
Click to view available options
Quantity:
1 g
5 g

CAS: 7101-31-7 | C2H6Se2 | 188.012 g/mol

Dimethyl diselenide is a powerful reagent used for identification of distonic radical cations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7101-31-7
Molecular Formula C2H6Se2
Molecular Weight (g/mol) 188.012
MDL Number MFCD00008254
InChI Key VLXBWPOEOIIREY-UHFFFAOYSA-N
Synonym dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci
PubChem CID 23496
IUPAC Name (methyldiselanyl)methane
SMILES C[Se][Se]C

Specifications

Density 1.987 g/mL
Boiling Point 155°C to 157°C
Flash Point 60°C (140°F)
Odor Unpleasant
Refractive Index 1.6388
Quantity 5 g
UN Number UN2929
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 187.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl diselenide
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H301-H331-H226-H373
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 230-410-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.