Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl fumarate, 99%

Catalog No. AAA1040230
Click to view available options
Quantity:
25 g
100 g
250 g

CAS: 624-49-7 | C6H8O4 | 144.126 g/mol

Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).

Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00064438
InChI Key LDCRTTXIJACKKU-ONEGZZNKSA-N
Synonym dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate
PubChem CID 637568
ChEBI CHEBI:76004
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point 102°C to 105°C
Density 1.37
Boiling Point 192°C to 193°C
Quantity 250 g
Beilstein 774590
Merck Index 14,4287
Solubility Information Soluble in water (1.6mg/ml at 20°C),methanol (30-36mg/ml),ethanol (10mg/ml at 25°C),DMSO (29mg/ml at 25°C),and DMF (∽12mg/ml).
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material Dimethyl fumarate
Hazard Category H312-H315-H317-H319-H335
Hazard Statement GHS H Statement
H312-H315-H319-H317-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 210-849-0
RTECSNumber EM6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.