Learn More
Dimethyl fumarate, 99%
Description
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing
Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).
Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.

Specifications
Specifications
Melting Point | 102°C to 105°C |
Density | 1.37 |
Boiling Point | 192°C to 193°C |
Quantity | 25 g |
Beilstein | 774590 |
Merck Index | 14,4287 |
Solubility Information | Soluble in water (1.6mg/ml at 20°C),methanol (30-36mg/ml),ethanol (10mg/ml at 25°C),DMSO (29mg/ml at 25°C),and DMF (∽12mg/ml). |
Formula Weight | 144.13 |
Percent Purity | 99% |
Chemical Name or Material | Dimethyl fumarate |
Safety and Handling
Hazard Category | H312-H315-H317-H319-H335 |
Hazard Statement | GHS H Statement H312-H315-H319-H317-H335 Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c |
EINECSNumber | 210-849-0 |
RTECSNumber | EM6125000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.