Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl iminodiacetate hydrochloride, 98%

Catalog No. AAB2572906
Click to view available options
Quantity:
5 g

CAS: 39987-25-2 | C6H12ClNO4 | 197.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39987-25-2
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.62
MDL Number MFCD00216724
InChI Key IIWYYIACSUPJCN-UHFFFAOYSA-N
Synonym dimethyl iminodiacetate hydrochloride, dimethyl 2,2'-azanediyldiacetate hydrochloride, methyl 2-2-methoxy-2-oxoethyl amino acetate hydrochloride, dimethyl iminodiacetic acid hydrochloride, iminodiacetic acid dimethyl ester hydrochloride, glycine, n-2-methoxy-2-oxoethyl-, methyl ester, hydrochloride, methyl 2-methoxycarbonyl methyl amino acetate, chloride, zlchem 797, acmc-1aeyr, iminodiacetic dimethyl ester hydrochloride
PubChem CID 2727727
IUPAC Name methyl 2-[(2-methoxy-2-oxoethyl)amino]acetate;hydrochloride
SMILES [H+].[Cl-].COC(=O)CNCC(=O)OC

Specifications

Melting Point ∼170°C
Quantity 5 g
Beilstein 3703236
Formula Weight 197.62
Percent Purity 98%
Chemical Name or Material Dimethyl iminodiacetate hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.