Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Indole-2,3-dicarboxylate 99%, Thermo Scientific™

Catalog No. AC408391000
Click to view available options
Quantity:
100 mg
Packaging:
Glass Bottle

Chemical Identifiers

CAS 54781-93-0
Molecular Formula C12H11NO4
Molecular Weight (g/mol) 233.223
MDL Number MFCD00082731
InChI Key JNRXSMOJWKDVHC-UHFFFAOYSA-N
Synonym dimethyl indole-2,3-dicarboxylate, 2,3-dimethyl 1h-indole-2,3-dicarboxylate, indole-2,3-dicarboxylic acid, dimethyl ester, 1h-indole-2,3-dicarboxylic acid dimethyl ester, 1h-indole-2,3-dicarboxylicacid, 2,3-dimethyl ester, 1h-indole-2,3-dicarboxylic acid, 2,3-dimethyl ester, methyl 2-methoxycarbonyl indole-3-carboxylate, 2,3-dicarbomethoxyindole, maybridge1_006201, dimethyl 2,3-indoledicarboxylate
PubChem CID 143197
IUPAC Name dimethyl 1H-indole-2,3-dicarboxylate
SMILES COC(=O)C1=C(NC2=CC=CC=C21)C(=O)OC

Specifications

Melting Point 112.0°C to 115.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 100 mg
Formula Weight 233.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dimethyl indole-2, 3-dicarboxylate, 99%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.