Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl isophthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
1459-93-4
C10H10O4
194.19
MFCD00008433
VNGOYPQMJFJDLV-UHFFFAOYSA-N
dimethyl isophthalate,dimethyl m-phthalate,isophthalic acid dimethyl ester,methyl isophthalate,1,3-benzenedicarboxylic acid, dimethyl ester,morflex 1129,dimethyl 1,3-benzenedicarboxylate,dimethylisophthalate,methyl 3-carbomethoxy benzoate,isophthalic acid, dimethyl ester
15088
dimethyl benzene-1,3-dicarboxylate
COC(=O)C1=CC(=CC=C1)C(=O)OC
1459-93-4
C10H10O4
194.19
MFCD00008433
VNGOYPQMJFJDLV-UHFFFAOYSA-N
dimethyl isophthalate,dimethyl m-phthalate,isophthalic acid dimethyl ester,methyl isophthalate,1,3-benzenedicarboxylic acid, dimethyl ester,morflex 1129,dimethyl 1,3-benzenedicarboxylate,dimethylisophthalate,methyl 3-carbomethoxy benzoate,isophthalic acid, dimethyl ester
15088
dimethyl benzene-1,3-dicarboxylate
COC(=O)C1=CC(=CC=C1)C(=O)OC
Boiling Point 124°C (16.0 hPa)
Viscosity 2.14 mPa.s (99°C)
Infrared Spectrum Authentic
Packaging Plastic bottle
Color White
Flash Point 138°C
Quantity 500g
Melting Point >66.0°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H4(CO2CH3)2
Solubility Information Solubility in water: insoluble
Formula Weight 194.19
Physical Form Flakes or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Dimethyl isophthalate, 99%
Show More Show Less

EINECSNumber : 215-951-9

RTECSNumber : NT2540000

TSCA : TSCA