Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl maleate

Catalog No. 2381981KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 g
1 kg

Catalog No. 23-819-81KG

Supplier: Sigma Aldrich 2381981KG

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethyl maleate (dmm) is a reactive dienophile and undergoes ultrasonic irradiation promoted diels-alder reaction with substituted furans
  • Mesoporous siliceous sba-15-supported cu catalyzed gas phase hydrogenolysis of dmm to 1,4-butanediol (bdo) has been reported
  • Aluminium chloride has been reported to accelerate the diels-alder reaction of dmm and anthracene
  • Dmm can be synthesized by the esterification of maleic anhydride with sulfuric acid and methanol

Specifications

CAS 624-48-6
Density 1.152 g/mL (at 25°C (literature))
Boiling Point 204°C to 205°C (lit.)
Molecular Formula C6H8O4
Refractive Index n20/D 1.441 (literature)
Linear Formula CH3OCOCH=CHCOOCH3
MDL Number MFCD00008459
Quantity 1 kg
Molecular Weight (g/mol) 144.13
Percent Purity 96%
Biological Activity Respiratory System, Skin
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.