Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl maleate, 96%

Catalog No. p-118266
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum

CAS: 624-48-6 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-48-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00008459
InChI Key LDCRTTXIJACKKU-ARJAWSKDSA-N
Synonym dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
PubChem CID 5271565
ChEBI CHEBI:35460
IUPAC Name dimethyl (Z)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point -17.5°C
Density 1.1500g/mL
Boiling Point 204.0°C to 205.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430
Linear Formula CH3O2CCH=CHCO2CH3
Quantity 250 mL
Beilstein 02,751
Specific Gravity 1.15
Solubility Information Solubility in water: 80g/L (25°C)
Viscosity 4 mPa.s (20°C)
Formula Weight 144.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Dimethyl maleate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
EINECSNumber 210-848-5
RTECSNumber EM6300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.