Learn More
Dimethyl maleate, 96%
Description
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.
Solubility
Miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
Specifications
| Melting Point | -19°C |
| Density | 1.151 |
| Boiling Point | 204°C to 205°C |
| Flash Point | 91°C (195°F) |
| Refractive Index | 1.442 |
| Quantity | 5000 g |
| UN Number | UN2810 |
| Beilstein | 471705 |
| Solubility Information | Miscible with water. |
| Formula Weight | 144.13 |
| Show More |
Safety and Handling
| Hazard Category | H302-H317-H319-H335-H373 |
| Hazard Statement | GHS H Statement H311-H318-H227-H302-H315-H335 Toxic in contact with skin. Causes serious eye damage. Combustible liquid. Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 210-848-5 |
| RTECSNumber | EM6300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.