Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl-n-octylchlorosilane 96.0+%, TCI America™
SDP

Catalog No. D1827100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1827100ML 100 mL
D182725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1827100ML Supplier TCI America Supplier No. D1827100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18162-84-0
Molecular Formula C10H23ClSi
Molecular Weight (g/mol) 206.829
MDL Number MFCD00000511
InChI Key DBKNGKYVNBJWHL-UHFFFAOYSA-N
Synonym n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc
PubChem CID 87482
IUPAC Name chloro-dimethyl-octylsilane
SMILES CCCCCCCC[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 220°C
Quantity 100 mL
UN Number 2987
Formula Weight 206.83
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl-n-octylchlorosilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.