Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl nitroterephthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
Packaging:
Plastic Bottle
5292-45-5
C10H9NO6
239.183
MFCD00008422
PAYWCKGMOYQZAW-UHFFFAOYSA-N
dimethyl 2-nitroterephthalate,dimethyl nitroterephthalate,terephthalic acid, nitro-, dimethyl ester,unii-59qlu09dz4,1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester,1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester,nitroterephthalic acid dimethyl ester,1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate,acmc-1aj3c,di-methyl nitroterephthalate
21364
dimethyl 2-nitrobenzene-1,4-dicarboxylate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)[N+](=O)[O-]
5292-45-5
C10H9NO6
239.183
MFCD00008422
PAYWCKGMOYQZAW-UHFFFAOYSA-N
dimethyl 2-nitroterephthalate,dimethyl nitroterephthalate,terephthalic acid, nitro-, dimethyl ester,unii-59qlu09dz4,1,4-benzenedicarboxylic acid, 2-nitro-, dimethyl ester,1,4-benzenedicarboxylic acid, 2-nitro-, 1,4-dimethyl ester,nitroterephthalic acid dimethyl ester,1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate,acmc-1aj3c,di-methyl nitroterephthalate
21364
dimethyl 2-nitrobenzene-1,4-dicarboxylate
COC(=O)C1=CC(=C(C=C1)C(=O)OC)[N+](=O)[O-]
Packaging Plastic Bottle
Color Yellow
Quantity 250g
Melting Point 73°C to 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Beilstein 09, 826
Formula Weight 239.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Dimethyl nitroterephthalate, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-134-1

TSCA : TSCA