Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dimethyl PDCD4 (Arg110) Mouse anti-Human, Clone: 30000000, MilliporeSigma™

Catalog No. MABC174625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC174625 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC174625 Supplier MilliporeSigma Supplier No. MABC174625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

dimethyl PDCD4 (Arg110) Monoclonal antibody specifically detects dimethyl PDCD4 (Arg110) in Human samples. It is validated for Immunocytochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dimethyl PDCD4 (Arg110)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 30000000
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal half of human Programmed cell death protein 4 with Arg 110 symmetrically dimethylated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.