Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Phenylphosphonate 98.0+%, TCI America™
SDP

Catalog No. p033225ml Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 2240-41-7
Molecular Formula C8H11O3P
Molecular Weight (g/mol) 186.147
MDL Number MFCD00059292
InChI Key OXDOANYFRLHSML-UHFFFAOYSA-N
Synonym Benzenephosphonic Acid Dimethyl Ester, Dimethyl Benzenephosphonate, Phenylphosphonic Acid Dimethyl Ester
PubChem CID 75234
IUPAC Name dimethoxyphosphorylbenzene
SMILES COP(=O)(C1=CC=CC=C1)OC

Specifications

Color Yellow
Boiling Point 247°C
Quantity 25 mL
Formula Weight 186.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Phenylphosphonate
EINECSNumber (3)-2521
RTECSNumber TA0378000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.