Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl phthalate, 99%

Catalog No. AAA142530E
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA142530E 2500 g
AAA1425330 250 g
AAA1425336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA142530E Supplier Thermo Scientific Chemicals Supplier No. A142530E
Add to Cart

CAS: 131-11-3 | C10H10O4 | 194.19 g/mol

Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

Solubility
Miscible with alcohol, ethanol, ether, benzene, chloroform, esters, ketones, organic solvents and chlorinated hydrocarbons. Slightly miscible with carbon tetrachloride. Immiscible with water, petroleum ether and paraffin hydrocarbons.

Notes
Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008425
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
PubChem CID 8554
ChEBI CHEBI:4609
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Melting Point ∼5°C
Density 1.19
Boiling Point 283°C to 284°C
Flash Point 146°C (294°F)
Assay Percent Range 99%
Refractive Index 1.515
Quantity 2500 g
Beilstein 1911460
Merck Index 14,3255
Solubility Information Miscible with alcohol,ethanol,ether,benzene,chloroform,esters,ketones,organic solvents and chlorinated hydrocarbons. Slightly miscible with carbon tetrachloride. Immiscible with water,petroleum ether and paraffin hydrocarbons.
IUPAC Name dimethyl benzene-1,2-dicarboxylate
Formula Weight 194.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl phthalate
Show More Show Less
EINECSNumber 205-011-6
RTECSNumber TI1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.