Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl pimelate, 98+%

Catalog No. AAB2202614
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1732-08-7 | C9H16O4 | 188.22 g/mol

Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

Solubility
Soluble in ethanol, benzene, ether. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1732-08-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00008470
InChI Key SHWINQXIGSEZAP-UHFFFAOYSA-N
Synonym dimethyl pimelate, heptanedioic acid, dimethyl ester, pimelic acid dimethyl ester, pimelic acid, dimethyl ester, dimethyl 1,7-heptanedioate, 1,7-dimethyl heptanedioate, heptanedioic acid, 1,7-dimethyl ester, unii-ge1pf3049f, heptanedioic acid dimethyl ester, 1ov5vo1
PubChem CID 74416
IUPAC Name dimethyl heptanedioate
SMILES COC(=O)CCCCCC(=O)OC

Specifications

Density 1.041
Boiling Point 121°C to 122°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Mild
Refractive Index 1.431
Linear Formula CH3O2C(CH2)5CO2CH3
Quantity 25 g
Beilstein 1777309
Solubility Information Soluble in ethanol,benzene,ether. Slightly soluble in water.
Formula Weight 188.23
Percent Purity 99%
Chemical Name or Material Dimethyl pimelate
Show More Show Less
EINECSNumber 217-057-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.