Learn More
Dimethyl pimelate, 98+%
Description
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.
Solubility
Soluble in ethanol, benzene, ether. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Specifications
| Density | 1.041 |
| Boiling Point | 121°C to 122°C (11 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Mild |
| Refractive Index | 1.431 |
| Linear Formula | CH3O2C(CH2)5CO2CH3 |
| Quantity | 5 g |
| Beilstein | 1777309 |
| Solubility Information | Soluble in ethanol,benzene,ether. Slightly soluble in water. |
| Formula Weight | 188.23 |
| Show More |
Safety and Handling
| EINECSNumber | 217-057-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.