Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl succinate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
106-65-0
C6H10O4
146.14
MFCD00008466
MUXOBHXGJLMRAB-UHFFFAOYSA-N
dimethyl succinate,methyl succinate,dimethylsuccinate,butanedioic acid, dimethyl ester,succinic acid, dimethyl ester,dbe-4 dibasic ester,methyl butanedioate,succinic acid dimethyl ester,butanedioic acid, 1,4-dimethyl ester,fema no. 2396
7820
dimethyl butanedioate
COC(=O)CCC(=O)OC
106-65-0
C6H10O4
146.14
MFCD00008466
MUXOBHXGJLMRAB-UHFFFAOYSA-N
dimethyl succinate,methyl succinate,dimethylsuccinate,butanedioic acid, dimethyl ester,succinic acid, dimethyl ester,dbe-4 dibasic ester,methyl butanedioate,succinic acid dimethyl ester,butanedioic acid, 1,4-dimethyl ester,fema no. 2396
7820
dimethyl butanedioate
COC(=O)CCC(=O)OC
Specific Gravity 1.11
Water 0.1% max. (K.F.)
Flash Point 85°C
Quantity 500g
Refractive Index 1.4185 to 1.4205
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.1100g/mL
Color Undesignated
Boiling Point 200.0°C
Acidity 0.2mg KOH/g max.
Melting Point 18.0°C to 19.0°C
Assay Percent Range 99%
Linear Formula CH3O2CCH2CH2CO2CH3
Beilstein 02, 609
Merck Index 15, 8999
Solubility Information Solubility in water: 75g/L (20°C)
Formula Weight 146.14
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Dimethyl succinate, 99%
Show More Show Less

Warning

EINECSNumber : 203-419-9

RTECSNumber : WM7675000

TSCA : TSCA