Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl succinate, 99%

Catalog No. p-120650
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 106-65-0 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-65-0
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00008466
InChI Key MUXOBHXGJLMRAB-UHFFFAOYSA-N
Synonym dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396
PubChem CID 7820
IUPAC Name dimethyl butanedioate
SMILES COC(=O)CCC(=O)OC

Specifications

Acidity 0.2mg KOH/g max.
Melting Point 18.0°C to 19.0°C
Density 1.1100g/mL
Boiling Point 200.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4185 to 1.4205
Linear Formula CH3O2CCH2CH2CO2CH3
Quantity 100 g
Beilstein 02, 609
Specific Gravity 1.11
Merck Index 15, 8999
Solubility Information Solubility in water: 75g/L (20°C)
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl succinate, 99%
Show More Show Less
Hazard Category Warning
EINECSNumber 203-419-9
RTECSNumber WM7675000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.