Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, 99+%

Catalog No. AAA1328036
Click to view available options
Quantity:
500 g
2500 g
10 kg

Catalog No. AAA1328036

Supplier: Thermo Scientific Chemicals A1328036

Only null left
Add to Cart

CAS: 67-68-5 | C2H6OS | 78.13 g/mol

Dimethyl Sulfoxide (DMSO) is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. It is also used in antifreeze or hydraulic fluids and utilized in the oxidation of thiols and disulfides to sulfonic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl Sulfoxide (DMSO) is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. It is also used in antifreeze or hydraulic fluids and utilized in the oxidation of thiols and disulfides to sulfonic acids. It is also used as a cryoprotectant and is added to cell media to reduce ice formation and thereby prevent cell death during the freezing process.

Solubility
Miscible with water, ethanol, ether, benzene, chloroform and acetone.

Notes
Hygroscopic. Incompatible with acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Density 1.101
Boiling Point 189°C
Quantity 500 g
Flash Point 87°C (189°F)
Odor Mild
Refractive Index 1.479
Linear Formula (CH3)2SO
Merck Index 143259
Solubility Information Miscible with water,ethanol,ether,benzene,chloroform and acetone.
Formula Weight 78.13
Percent Purity ≥99%
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P370+P378q-P501c
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.